Cytiva HyClone™ Minimum Essential Medium
Support cell culture growth and biomanufacturing processes with HyClone™ Minimum Essential Medium (MEM) with Earle's Balanced Salts (MEM/EBSS), manufactured based on compliance with ISO 9001:2000 guidelines and current Good Manufacturing Practices (cGMP) for Medical Devices.
Supplier:  Cytiva SH3026502
 View more versions of this product
Catalog No. SH3026502
Explore available promotions
Description
Description
  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured using ISO 9001 certified processes
Specifications
Specifications
cGMP, ISO 9001:2000
MEM Alpha Modification, with L-Glutamine, with Ribo- and Deoxyribonucleosides
MEM Alpha modification, with L-glutamine, ribo- and deoxyribonucleosides
0.1 μm Sterile-filtered
None
2° to 8°C, away from light
Liquid
Sterile-filtered
Glutamine, Ribonucleosides, Deoxyribonucleosides
1000 mL