missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ RPMI 1640 Media

Designed to support cell culture growth and biomanufacturing processes.

$13.17 - $129.13

Specifications

Certifications/Compliance ISO 9001:2000
Content And Storage 2° to 8°C, away from light
Form Liquid
Product Type RPMI 1640
Sterilization Method 0.1 μm Sterile-filtered
View More Specs

Products 12
Catalog Number Mfr. No. Quantity With Additives Price Quantity  
Catalog Number Mfr. No. Quantity With Additives Price Quantity  
SH30027FS
View Documents Promotion Details
Cytiva
SH30027FS
6 x 500 mL Glutamine, Phenol Red
Pack of 6 Each for $78.04
 
SH3002702
View Documents
Cytiva
SH3002702
1000 mL Glutamine, Phenol Red
Each for $21.88
 
SH30027LS
View Documents
Cytiva
SH30027LS
6 x 1000 mL Glutamine, Phenol Red
Pack of 6 Each for $108.38
 
SH3009602
View Documents
Cytiva
SH3009602
1000 mL Phenol Red
Each for $23.98
 
SH30096LS
View Documents
Cytiva
SH30096LS
6 x 1000 mL Phenol Red
Pack of 6 Each for $129.13
 
SH30096FS
View Documents
Cytiva
SH30096FS
6 x 500 mL Phenol Red
Pack of 6 Each for $83.46
 
SH30255FS
View Documents
Cytiva
SH30255FS
6 x 500 mL Glutamine, HEPES, Phenol Red
Pack of 6 Each for $112.04
 
SH3025501
View Documents
Cytiva
SH3025501
500 mL Glutamine, HEPES, Phenol Red
Each for $19.17
 
SH3025502
View Documents
Cytiva
SH3025502
1000 mL HEPES, Phenol Red, Phenol Red
Each for $32.38
 
SH3060501
View Documents
Cytiva
SH3060501
500 mL
Each for $21.71
 
SH3009601
View Documents Promotion Details
Cytiva
SH3009601
500 mL Phenol Red
Each for $14.65
 
SH3002701 Edge
View Documents Promotion Details
Cytiva
SH3002701
500 mL Glutamine, Phenol Red
Each for $13.17
Edge
 
Description

Description

  • Format: Liquid
Specifications

Specifications

ISO 9001:2000
Liquid
0.1 μm Sterile-filtered
2° to 8°C, away from light
RPMI 1640
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'