missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Description
Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.
Specifications
cGMP | |
SFM4HEK293 Media | |
0.2 μm Sterile-filtered | |
No Protein, No Phenol Red, No HEPES |
2° to 8°C, away from light | |
Liquid | |
Glutamine, Poloxamer 188, Sodium Bicarbonate | |
1000 mL |