missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Water for Injection (WFI) Quality Water
Supports cell culture growth and biomanufacturing process.
$660.88 - $3149.90
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
SH3022124
|
Cytiva
SH3022124 |
10 L |
Each for $660.88
|
|
||||||
SH3022125
|
Cytiva
SH3022125 |
20 L |
Each for $722.14
|
|
||||||
SH3022126
|
Cytiva
SH3022126 |
50 L |
Each for $1,572.82
|
|
||||||
SH3022127
|
Cytiva
SH3022127 |
100 L |
Each for $2,702.71
|
|
||||||
SH3022128
|
Cytiva
SH3022128 |
200 L |
Each for $3,149.90
|
|
||||||
Description
- Format: Liquid
Chemical Identifiers
7732-18-5 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
H2O | |
dihydrogen oxide, dihydrogen monoxide |
Specifications
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.02 | |
CHEBI:15377 | |
Water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
962 | |
10 L |
Safety and Handling
ShelfLife : 36 Months