missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HyClone™ Water, Molecular Biology Grade, Cytiva Edge
Edge

For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.

Supplier:  Cytiva SH3053801

 View more versions of this product

Catalog No. SH3053801


Edge
Explore available promotions

Description

Description

  • Format: Liquid
Specifications

Specifications

100 mL

Chemical Identifiers

0.0
Safety and Handling

Safety and Handling

ShelfLife : 24 Months

Recommended Storage : 2° to 30°C

SDS
Documents

Documents

Promotions

Promotions