HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™
Supplier:  Honeywell Inc 348166X1LUS
 View more versions of this product
Catalog No. 60-046-468
Explore available promotions
Description
Description
Specifications
Specifications
∼1.15 g/mL
6 x 1 L
Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)
≥70.0000%,≥5.0000%,≥5.0000%,≥1.0000%,≥1.0000%,≥1.0000%
Glass Bottle
Karl Fischer
99°C
HYDRANAL™ - Composite 5 K
111-90-0,7553-56-2,288-32-4,693-98-1,7446-09-5,68007-08-9
<90.0000%,<10.0000%,<10.0000%,<5.0000%,<5.0000%,<5.0000%
DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol