missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Coulomat A, Reagent for coulometric KF titration (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™
$249.88 - $249.88
Specifications
Density | 1.070 g/cm3 |
---|---|
Flash Point | 11°C |
Chemical Name or Material | HYDRANAL™ - Coulomat A |
Name Note | Reagent for coulometric KF titration (anolyte solution), preferred for cells with diaphragm |
CAS Min % | ≥30.0000%,≥30.0000%,≥10.0000%,≥5.0000%,≥1.0000% |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60-046-449
|
Honeywell Inc
348076X500MLUS |
500 mL | Glass Bottle |
Each for $249.88
Case of 6 Each for $1,499.28
|
|
||||
Specifications
1.070 g/cm3 | |
HYDRANAL™ - Coulomat A | |
≥30.0000%,≥30.0000%,≥10.0000%,≥5.0000%,≥1.0000% | |
67-66-3,288-32-4,7446-09-5,68007-08-9 | |
Glass Bottle | |
Methyl alcohol |
11°C | |
Reagent for coulometric KF titration (anolyte solution), preferred for cells with diaphragm | |
<50.0000%,<50.0000%,<20.0000%,<10.0000%,<5.0000% | |
59°C | |
UN1992 | |
Coulometric KF Titration |