HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™
Supplier: Honeywell-Hydranal 348406X50ML
Specifications
1.064 g/cm3 | |
1 Kit | |
Reagent for coulometric KF titration (catholyte solution) | |
≥50.0000%,≥30.0000%,≥1.0000%,≥1.0000% | |
63°C | |
UN1230 |
14°C | |
HYDRANAL™ - Coulomat CG | |
14426-21-2,67-56-1,134227-25-1,7446-09-5 | |
<70.0000%,<50.0000%,≤5.0000%,<5.0000% | |
Ampoule | |
Karl Fischer |