missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Coulomat E, Reagent for coulometric KF titration (ethanol-based), for cells with and without diaphragm, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration (ethanol-based), for cells with and without diaphragm
Supplier: Honeywell-Fluka 347266X500ML
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
0.980 g/cm3 | |
6 x 500 mL | |
Reagent for coulometric KF titration (ethanol-based), for cells with and without diaphragm | |
<70.0000%,<20.0000%,<10.0000%,<10.0000%,<10.0000%,<10.0000% | |
67°C | |
UN1993 | |
Karl Fischer |
12°C | |
HYDRANAL™ - Coulomat E | |
≥50.0000%,≥10.0000%,≥5.0000%,≥5.0000%,≥5.0000%,≥5.0000% | |
111-42-2,67-56-1,101023-55-6,7446-09-5,68007-08-9 | |
Glass Bottle | |
Ethyl alcohol |