missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™
Supplier: Honeywell Inc 347414X25LUS
Specifications
11°C | |
∼0.79Â g/mL | |
Reagent for volumetric one-component KF titration (working medium) | |
64°C to 65°C | |
CH4O | |
Methyl alcohol | |
CO | |
32.04 | |
Karl Fischer |
4 x 2.5 L | |
HYDRANAL™ - Methanol dry | |
≤100.0000% | |
Glass Bottle | |
MFCD00004595 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
32.04 |