missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™
Supplier: Honeywell-Hydranal 348016X500MLUS
Specifications
11°C | |
∼0.85Â g/mL | |
Reagent for volumetric two-component Karl Fischer titration | |
≤100.0000%,<10.0000% | |
65°C | |
UN1230 | |
Karl Fischer |
6 x 500 mL | |
HYDRANAL™ - Titrant 5 | |
≥90.0000%,≥5.0000% | |
7553-56-2 | |
Glass Bottle | |
Methyl alcohol |