missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Honeywell™ Fluka™
$251.38 - $251.38
Chemical Identifiers
CAS | 108-32-7 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008519 |
InChI Key | CTQNGGLPUBDAKN-UHFFFAOYSA-N |
Synonym | 1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | 1,2-xylene |
SMILES | CC1=CC=CC=C1C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
60050009
|
Honeywell Chemicals
348496X80ML |
6 x (10 x 8 mL) |
Each for $251.38
Case of 6 Each for $1,508.28
|
|
||||||
Chemical Identifiers
108-32-7 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
962 | |
1,2-xylene |
C8H10 | |
MFCD00008519 | |
1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one | |
CHEBI:15377 | |
CC1=CC=CC=C1C |
Specifications
1.00Â g/mLÂ at 20Â °C | |
6 x (10 x 8 mL) | |
≥50.0000%,≥20.0000%,≥20.0000% | |
109°C | |
C8H10 | |
UN1993 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
962 | |
Karl Fischer | |
effective water |
32°C | |
108-32-7,95-47-6,71-36-3 | |
<70.0000%,<30.0000%,<30.0000% | |
Ampoule | |
MFCD00008519 | |
1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one | |
CC1=CC=CC=C1C | |
106.17 | |
CHEBI:15377 | |
HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), exact value on report of analysis |
Safety and Handling
ShelfLife : 1800 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Flammable liquids, n.o.s.
Recommended Storage : Room Temp