Learn More
Hydrazine acetate, 97%
CAS: 7335-65-1 | C2H8N2O2 | 92.10 g/mol
$141.40 - $466.54
Chemical Identifiers
| CAS | 7335-65-1 |
|---|---|
| Molecular Formula | C2H8N2O2 |
| Molecular Weight (g/mol) | 92.10 |
| MDL Number | MFCD00013141 |
| InChI Key | YFHNDHXQDJQEEE-UHFFFAOYSA-N |
| Synonym | hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph |
| PubChem CID | 165591 |
| IUPAC Name | acetic acid;hydrazine |
| SMILES | NN.CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC411750050
|
Thermo Scientific Chemicals
411750050 |
5 g | Glass bottle |
Each for $141.40
|
|
||||
|
AC411750250
|
Thermo Scientific Chemicals
411750250 |
25 g | Glass bottle |
Each for $466.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
| 7335-65-1 | |
| 92.10 | |
| YFHNDHXQDJQEEE-UHFFFAOYSA-N | |
| 165591 | |
| NN.CC(O)=O |
| C2H8N2O2 | |
| MFCD00013141 | |
| hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph | |
| acetic acid;hydrazine |
Specifications
| 7335-65-1 | |
| Authentic | |
| Glass bottle | |
| NH2NH2·CH3COOH | |
| 5 g | |
| YFHNDHXQDJQEEE-UHFFFAOYSA-N | |
| acetic acid;hydrazine | |
| 165591 | |
| 97% | |
| Hydrazine acetate, 99% |
| White | |
| 97% | |
| C2H8N2O2 | |
| MFCD00013141 | |
| hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph | |
| NN.CC(O)=O | |
| 92.10 | |
| 92.1 | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
May cause an allergic skin reaction.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN: Gently wash with plenty of soap and water
GHS Signal Word: Danger
EINECSNumber : 230-845-2
TSCA : TSCA
RUO – Research Use Only