missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate, 55% (Hydrazine, 35%)

CAS: 10217-52-4 | H4N2 | 32.05 g/mol

$177.67 - $771.34

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES NN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296811000
SDS View Documents
Thermo Scientific Chemicals
296811000
100 mL Glass bottle
Each for $177.67
Only null left
 
AC296815000
SDS View Documents
Thermo Scientific Chemicals
296815000
500 mL Glass bottle
Each for $192.08
Only null left
 
AC296810025
View Documents
Thermo Scientific Chemicals
296810025
2.5 L Glass bottle
Each for $771.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

10217-52-4
32.05
OAKJQQAXSVQMHS-UHFFFAOYSA-N
24654
NN
H4N2
MFCD00149931
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
CHEBI:35511

Specifications

109.4°C
10217-52-4,7732-18-5
100 mL
H4N2
MFCD00149931
>100°C
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
OAKJQQAXSVQMHS-UHFFFAOYSA-N
hydrazine;hydrate
24654
32.04
Glass bottle
1.023
Hydrazine hydrate, 55% (Hydrazine 35%)
-65.0°C
Liquid
55%
H2NNH2·xH2O
01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
14,4771
Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether
NN
32.05
CHEBI:35511
55% (Hydrazine,35%)
1.3870 to 1.3910
1.0230g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
May cause cancer.
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Causes severe skin b

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m

GHS Signal Word: Danger

RTECSNumber : MV4590000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only