missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Hydrazinocarbonyl)ferrocene 95.0+%, TCI America™
Supplier: TCI America H09411G
Specifications
(Hydrazinocarbonyl)ferrocene [for HPLC Labeling] | |
C11H12FeN2O | |
1 g | |
244.08 | |
Crystalline Powder |
12153-28-5 | |
MFCD01321175 | |
Ferrocenecarbohydrazide | |
≥95.0% (HPLC,T) |
Chemical Identifiers
12153-28-5 | |
C11H12FeN2O | |
MFCD01321175 | |
Ferrocenecarbohydrazide |
Safety and Handling
TSCA : No