missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Micro Essential Lab Hydrion™ Urine and Saliva pH Paper, Range 5.5-8.0

Provides excellent color separation in both the acid range and the alkali range

Supplier:  Micro Essential Lab MES067

Catalog No. 14-853-275


Explore available promotions

Description

Description

  • Commonly used to test urine and saliva, it offers an easy to read, single color match at every 0.2 to 0.3 interval from pH 5.5-8.0
  • Includes color chart

Specifications

Specifications

pH, Urine
Urinalysis
Color chart
100 Tests
Videos
SDS
Documents

Documents

Promotions

Promotions