Learn More
Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid.
Supplier: Thermo Scientific™ TS42801
Alert:
Material is flammable before film is formed and HCl fumes are generated in its application. Fluid is acidic during application and care should be taken to avoid corrosion of metal that comes into contact with the liquid. After application, the surface is neutral.
Description
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid. This short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface. It is excellent for modifying metals, certain plastics, ceramics and fiber optics.
Additional Product InformationHydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.
The fluid is acidic only during application; after application, the surface is neutral.
Specifications
Siliconizing Glass Surfaces | |
98 to 100% | |
480 mL | |
Hydrocarbon Soluble Siliconizing Fluid |
1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane | |
480 mL | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.