missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.0 N Aqueous Solution, APHA, 1 N +/-0.005 N, Spectrum™ Chemical
H-135, 7647-01-0
$157.77 - $1772.20
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603167
|
Spectrum Chemical Mfg Cor
H1351LT |
1 L | Poly Bottle |
Each for $157.77
|
|
||||
|
18603168
|
Spectrum Chemical Mfg Cor
H1354LTPL |
4 L | Poly Bottle |
Each for $292.37
|
|
||||
|
18603170
|
Spectrum Chemical Mfg Cor
H1354LTPLCS4 |
4 x 4 L | Poly Bottle |
Case of 4 Each for $885.07
|
|
||||
|
18603169
|
Spectrum Chemical Mfg Cor
H13520LT |
20 L | Poly Bottle |
Each for $689.17
|
|
||||
|
18603171
|
Spectrum Chemical Mfg Cor
H135200LTBL |
200 L | Poly Bottle |
Each for $1,772.20
|
|
||||
Description
Spectrum™ Chemical Hydrochloric Acid, 1.0 N Aqueous Solution, APHA , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |
Specifications
| 7647-01-0,7732-18-5 | |
| 3.6 to 3.7%,96.3 to 96.4% | |
| 1 L | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| Ungraded |
| 1 N ±0.005 | |
| Poly Bottle | |
| ClH | |
| UN1789 | |
| Cl | |
| 36.46 | |
| Hydrochloric Acid |