missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 10% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company 357025

 View more versions of this product

Catalog No. 35702-5


Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7647-01-0,7732-18-5
10% (v/v)
Cubitainer
10 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Colorless
88.0,10.34
91.0,10.77
Aqueous Solution
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Videos
SDS Documents

Documents

Promotions

Promotions