missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 50% (v/v), Aqueous Solution, APHA, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H19520LT

 View more versions of this product

Catalog No. 18610922



Description

Description

Specifications

Specifications

7647-01-0,7732-18-5
Cubitainer
1.09 - 1.11
20 L

Chemical Identifiers

7647-01-0
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.