missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, ACS, Fisher Chemical
Supplier: Thermo Fisher Scientific Formerly Fairlawn A149C4
Specifications
Hydrochloric Acid | |
7647-01-0 | |
4 L | |
Liquid | |
palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440 | |
Cl | |
36.46 | |
CHEBI:53434 | |
36.5 to 38.0% |
Colorless | |
1ppm max. | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
24290 | |
36.4600 | |
ACS |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
24290 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440 | |
CHEBI:53434 | |
Cl |