missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, ACS Grade, 36.5 to 38.0%, LabChem™
Supplier: LabChem LC1495036
Specifications
Hydrochloric Acid | |
7647-01-0,7732-18-5 | |
63,37 | |
HDPE Bottle | |
Hydrogen chloride | |
ClH | |
HCl | |
1789 | |
Soluble in water | |
Cl | |
36.46 | |
CHEBI:17883 | |
36.5 to 38.0% |
Colorless | |
1190kg/m3 | |
Passes Test | |
1190kg/m3 | |
6 x 2.5 L | |
Liquid | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
ACS |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life.
GHS P Statement
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Us
Danger
DOTInformation : Class 8
EINECSNumber : 231-595-7
Recommended Storage : Room Temperature