Learn More
Hydrochloric Acid, ACS Grade, 36.5 to 38.0%, LabChem™
Supplier: LabChem LC1495036S
Specifications
Hydrochloric Acid | |
7647-01-0,7732-18-5 | |
63,37 | |
Amber Glass | |
Hydrogen chloride | |
ClH | |
HCl | |
UN1789 | |
Soluble in water | |
Cl | |
36.46 | |
CHEBI:17883 | |
36.5 to 38.0% |
Colorless | |
1,190kg/m3 | |
Passes Test | |
1190kg/m3 | |
6 x 2.5 L | |
Liquid | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
ACS |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life.
GHS P Statement
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with applicable regulations.
Danger
DOTInformation : ass 8
EINECSNumber : 231-595-7
Recommended Storage : Room Temperature