missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, ACS Grade, 36.5 to 38.0%, LabChem™

Supplier:  LabChem LC149503S

 View more versions of this product

Catalog No. LC149503S


Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7647-01-0,7732-18-5
63,37
PVC Coated
Hydrogen chloride
ClH
HCl
UN1789
Soluble in water
Cl
36.46
CHEBI:17883
36.5 to 38.0%
Colorless
1,190kg/m3
Passes Test
1190kg/m3
2.5 L
Liquid
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
ACS

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with applicable regulations.

Danger

DOTInformation : ass 8

EINECSNumber : 231-595-7

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions