missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™

Supplier:  Thermo Fisher Scientific Formerly Fairlawn FLA144500LB

 View more versions of this product

Catalog No. A144-500LB


Explore available promotions

Description

Description

Specifications

Specifications

<1
-35°C
57°C
0.1ppm max.
Poly Drum
1.18
500 lb.
Liquid
Soluble in water
Cl
36.46
Colorless
7647-01-0
1.18g/cm³
Pass Test
4ppm max.
125mbar at 20°C
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36.5 to 38.0% (w/w)

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:1

SDS Documents

Documents

Accessories
Promotions

Promotions