Learn More
Hydrochloric acid, pure, fuming, 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, 35.0 to 39.0%, ClH, CAS Number-7647-01-0, 7732-18-5, acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 57.0 deg.C, CHEBI:17883
$82.99 - $164.93
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC124620010
|
Thermo Scientific Chemicals
124620010 |
1 L | Glass Bottle |
Each for $82.99
Case of 4 Each for $0.00
|
|
||||
AC124620025
|
Thermo Scientific Chemicals
124620025 |
2.5 L | Glass Bottle |
Each for $142.85
Case of 4 Each for $0.00
|
|
||||
AC124620026
|
Thermo Scientific Chemicals
124620026 |
2.5 L | Plastic Bottle |
Each for $164.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0 | |
36.46 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
Specifications
-35.0°C | |
57.0°C | |
Glass Bottle | |
1 L | |
ClH | |
HCl | |
04,450; 05,533; 06,283 | |
Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons | |
Cl | |
36.46 | |
CHEBI:17883 | |
35.0 to 39.0% | |
Hydrochloric acid |
7647-01-0,7732-18-5 | |
1.1800g/mL | |
1.18 | |
1.9 mPa.s (15°C) | |
Solution | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 | |
Pure |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only