missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid Solution, 0.1N (N/10) (Certified), Fisher Chemical™

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific SA5410

 View more versions of this product

Catalog No. SA54-10


Explore available promotions

Description

Description

Specifications

Specifications

0.10
7647-01-0,7732-18-5
Pass Test
LOT 84L Potassium acid phthalate
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
0.1N (N/10)
Hydrochloric Acid Solution, 0.1N (N/10)
0.0995 to 0.1005N
PolyPac™
10 L
Liquid
1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Certified

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word: CAUTION!

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions