missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, 0.2M, Honeywell Fluka™
Volumetric, 0.2Â M HCl (0.2N)
Supplier: Honeywell-Fluka 3431022L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
| Hydrochloric acid solution | |
| 0.2M | |
| 190 mmHg (25°C) | |
| ClH | |
| UN1789 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| chlorane | |
| 313 | |
| 36.46g/mol |
| 7647-01-0 | |
| 1.005g/mL at 25°C | |
| 2 L | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |