missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid Solution, 1N (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Supplier: Thermo Fisher Scientific FLSA4820
Specifications
1.1 | |
0°C | |
100°C | |
Pass Test | |
LOT 84L Potassium acid phthalate | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
Certified |
Hydrochloric Acid Solution, 1N | |
7647-01-0,7732-18-5 | |
0.995 to 1.005N | |
PolyPac™ | |
20 L | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
1N |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Safety and Handling
CAUTION!
Emergency Overview
Corrosive to metals. Irritating to eyes and skin. Harmful by inhalation, in contact with skin and if swallowed.
NFPA
Health:
Signal Word: DANGER!
DOTInformation : DOT Class 8, : Corrosive