missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
$448.83 - $1068.00
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A508-P500
|
Seastar Chemicals Inc
S010401TFIQ03 |
500 mL | HDPE Bottle |
Each for $448.83
|
|
||||
A508-P212
![]() |
Seastar Chemicals Inc
S010401TFIQ09 |
2.5 L | HDPE Bottle |
Each for $927.33
|
![]() |
||||
A5084
|
Seastar Chemicals Inc
S010401TFIH43 |
4 L | HDPE Bottle |
Each for $1,068.00
Case of 4 Each for $4,272.00
|
|
||||
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
1 | |
-35°C | |
57°C | |
HDPE Bottle | |
Certificate of Lot Analysis included with each shipment. | |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
TraceMetal |
Colorless | |
7647-01-0 | |
1.18g/cm³ | |
125mbar at 20°C | |
500 mL | |
Liquid | |
1789 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
Hydrochloric Acid |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
ShelfLife : 3 years
DOTInformation : DOT Class 8, : Corrosive