Learn More
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
$83.12 - $83.12
Chemical Identifiers
CAS | 501-52-0 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD00002771 |
InChI Key | XMIIGOLPHOKFCH-UHFFFAOYSA-N |
Synonym | hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid |
PubChem CID | 107 |
ChEBI | CHEBI:28631 |
IUPAC Name | 3-phenylpropanoic acid |
SMILES | C1=CC=C(C=C1)CCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC159101000
|
Thermo Scientific Chemicals
159101000 |
100 g | Glass bottle |
Each for $83.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
501-52-0 | |
150.18 | |
XMIIGOLPHOKFCH-UHFFFAOYSA-N | |
107 | |
3-phenylpropanoic acid |
C9H10O2 | |
MFCD00002771 | |
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid | |
CHEBI:28631 | |
C1=CC=C(C=C1)CCC(=O)O |
Specifications
501-52-0 | |
1.0700g/mL | |
>112°C | |
98.5% min. (GC) | |
C9H10O2 | |
MFCD00002771 | |
09, 508 | |
15, 4822 | |
Solubility in water: 0.5% in water; more soluble in hot water. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,benzene,ether,chloroform,,petroleum ether and carbon disulfide | |
C1=CC=C(C=C1)CCC(=O)O | |
150.18 | |
CHEBI:28631 | |
99% | |
Hydrocinnamic acid, 99% |
47.0°C to 50.0°C | |
279.0°C to 281.0°C | |
Authentic | |
Glass bottle | |
C6H5CH2CH2CO2H | |
100 g | |
1.07 | |
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid | |
XMIIGOLPHOKFCH-UHFFFAOYSA-N | |
3-phenylpropanoic acid | |
107 | |
150.18 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 207-924-5
RTECSNumber : DA8600000
TSCA : TSCA
RUO – Research Use Only