missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Bromide, 33% Solution in Acetic Acid, 31 -35%, Spectrum™ Chemical
$936.53 - $82408.59
Specifications
| CAS | 64-19-7,10035-10-6 |
|---|---|
| Percent Purity | 31 to 35% |
| Grade | Reagent |
| Appearance | Light to Dark Yellow Liquid |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610942
|
Spectrum Chemical Mfg Cor
H1141500MLGL |
500 mL | Amber Glass Bottle |
Each for $936.53
|
|
||||
|
18610943
|
Spectrum Chemical Mfg Cor
H11411LTGL |
1 L | Amber Glass Bottle |
Each for $1,493.00
|
|
||||
|
18610944
|
Spectrum Chemical Mfg Cor
H1141100KGBL |
100 kg | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| 64-19-7,10035-10-6 | |
| 31 to 35% | |
| Reagent | |
| Light to Dark Yellow Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.