missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, nominally 2.5M in ethanol

CAS: 7647-01-0 | ClH | 36.46 g/mol

$254.21 - $847.34

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44605AK
View Documents
Thermo Scientific Chemicals
044605AK
250 mL
Each for $254.21
Only null left
 
AA44605K2
View Documents
Thermo Scientific Chemicals
044605K2
1 L
Each for $847.34
Only null left
 
Description

Description

Hydrogen chloride, 2.5M. in ethanol is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883

Specifications

7647-01-0
MFCD00011324 MFCD00792839
Nominally 2.5M in ethanol
Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
Cl
36.46
CHEBI:17883
250 mL
Hydrochloric acid
ClH
UN2924
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.

P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c

H225-H290-H314-H332-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only