missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 30% w/w, ACS Guaranteed Reagent), MilliporeSigma™

Supplier:  Technidata HX06352

Catalog No. MHX06352

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Hydrogen Peroxide, 29.0–32.0%
-33°C
108°C
H2O2
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
ACS
7722-84-1
1.11kg/L
Plastic Bottle
1 L
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
CHEBI:16240

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
hydrogen peroxide
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
CHEBI:16240
OO
Videos
SDS
Documents

Documents

Promotions

Promotions