missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, 3% Solution, Reagent, 3.0-3.5%, Spectrum™ Chemical
$161.47 - $12694.85
Specifications
CAS | 7722-84-1,7732-18-5 |
---|---|
Percent Purity | 3.0 to 3.5% |
Grade | Reagent |
Specific Gravity | 1.01 |
Physical Form | Chemical Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610947
|
Spectrum Chemical Mfg Cor
H1060200LTBL |
200 L | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18610948
|
Spectrum Chemical Mfg Cor
H10601LT |
1 L | Poly Bottle |
Each for $258.65
|
|
||||
18610949
|
Spectrum Chemical Mfg Cor
H10604LT |
4 L | Poly Bottle |
Each for $510.91
|
|
||||
18610950
|
Spectrum Chemical Mfg Cor
H1060500MLPL |
500 mL | Poly Bottle |
Each for $161.47
|
|
||||
Specifications
7722-84-1,7732-18-5 | |
Reagent | |
Chemical Solution |
3.0 to 3.5% | |
1.01 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.