missing translation for 'onlineSavingsMsg'
Learn More

hydrogen peroxide, 3 wt.% solution in water, stabilized

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

$88.72 - $232.38

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426000250
SDS View Documents
Thermo Scientific Chemicals
426000250
25 mL Plastic bottle
Each for $88.72
Only null left
 
AC426001000
SDS View Documents
Thermo Scientific Chemicals
426001000
100 mL Plastic bottle
Each for $110.95
Only null left
 
AC426000010
SDS View Documents
Thermo Scientific Chemicals
426000010
1 L Plastic bottle
Each for $232.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7722-84-1
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
H2O2
MFCD00011333
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO

Specifications

7722-84-1,7732-18-5
H2O2
MFCD00011333
15, 4835
Solubility in water: soluble.
OO
34.014
CHEBI:16240
Plastic bottle
25 mL
100.0°C
HOOH
1.0100g/mL
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34
1.01
Hydrogen peroxide, 3 wt.% solution in water, stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only