missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
$165.06 - $12045.17
Chemical Identifiers
CAS | 7722-84-1 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
MDL Number | MFCD00011333 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
H325-500
|
Fisher Chemical
H325500 |
500 mL | Poly Bottle |
Each for $165.06
Case of 6 Each for $990.36
|
|
||||
H325-4
|
Thermo Fisher Scientific
H3254 |
4 L | Poly Bottle |
N/A
|
N/A | ||||
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
H3-253-0GAL
|
Fisher Chemical
H32530GAL |
30 gal. | Poly Drum |
N/A
|
N/A | ||||
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
Hydrogen Peroxide, 30% (Certified ACS)
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
Colorless | |
108°C | |
29.0 to 32.0 % | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
29.0 to 32.0% | |
Pass test | |
0.002 % max. | |
Decomposes information: >125°C | |
500 mL | |
Hydrogen Peroxide, 30% |
7722-84-1,7732-18-5 | |
-33°C | |
H2O2 | |
1.110g/cm³ | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34 | |
ACS | |
Poly Bottle | |
23mmHg at 30°C | |
3.3 | |
Liquid |
Safety and Handling
DANGER!
Emergency Overview
Causes severe eye irritation and possible burns. Irritating to skin. Harmful if swallowed.
NFPA
Health:3 Flammability:0 Instability:1
Signal Word: DANGER!
DOTInformation : DOT Class 5.1, : Oxidizer