missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, 30% (w/w), ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company 382171
Specifications
Hydrogen Peroxide | |
7722-84-1,7732-18-5 | |
35.0,75.0 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
30% (w/w) | |
4 L |
Colorless | |
25.0,65.0 | |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
ACS Reagent | |
Vented Poly Bottle | |
Liquid |
Chemical Identifiers
7722-84-1 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |