missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 30% (w/w), ACS Reagent Grade, Ricca Chemical

Supplier:  Ricca Chemical Company 382175

 View more versions of this product

Catalog No. 38217-5

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Hydrogen Peroxide
7722-84-1,7732-18-5
35.0,75.0
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
CHEBI:16240
30% (w/w)
20 L
Colorless
25.0,65.0
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
ACS Reagent
Cubitainer
Liquid

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
Videos
SDS Documents

Documents

Promotions

Promotions