missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 35% Solution, FCC, Spectrum™ Chemical
HY115, 7722-84-1, H2O2
$611.50 - $17029.87
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.01 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| IUPAC Name | peroxol |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603263
|
Spectrum Chemical Mfg Cor
HY115500MLPL |
500 mL | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
18603264
|
Spectrum Chemical Mfg Cor
HY115500MLPLCS6 |
6 x 500 mL | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
18603265
|
Spectrum Chemical Mfg Cor
HY1154LTPL |
4 L | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
18603266
|
Spectrum Chemical Mfg Cor
HY1154LTPLCS4 |
4 x 4 L | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
18603267
|
Spectrum Chemical Mfg Cor
HY115200LTBL |
200 L | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
Spectrum™ Chemical Hydrogen Peroxide, 35 Percent Solution, FCC is used as a disinfectant in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| 0.03% | |
| 65%,35% | |
| UN2014 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| peroxol | |
| 35% | |
| Poly Bottle | |
| Hydrogen Peroxide |
| 7732-18-5,7722-84-1 | |
| H2O2 | |
| 35% | |
| OO | |
| 34.01 | |
| FCC | |
| 500 mL |