missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 35% Solution, Technical, 30-36%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H10754LT

 View more versions of this product

Catalog No. 18610962

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

7722-84-1,7732-18-5
30 to 36%
Poly Bottle
1
Technical
4 L

Chemical Identifiers

7722-84-1
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.