missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Certified, 3.0% (w/w) to 3.5% (w/w), LabChem™
$54.70 - $213.50
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.014 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Specifications
| 7722-84-1,7732-18-5 | |
| 97,3 | |
| 500 mL | |
| H2O2 | |
| Soluble in water | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| 3.0 to 3.5% (w/w) | |
| Passes Test | |
| Oxygen | |
| Hydrogen Peroxide |
| Colorless | |
| Liquid | |
| H2O2 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01 | |
| Certified | |
| Poly Bottle | |
| 1g/mL |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Danger
Recommended Storage : Room Temperature