missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide Solution, 29.0-32.0%, ACS, MilliporeSigma™
Supplier: Technidata HX06356
Specifications
Hydrogen Peroxide Solution | |
7722-84-1 | |
29.0 to 32.0 % | |
H2O2 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
Liquid |
For ACS analysis | |
Colorless | |
Plastic Bottle | |
2.5 L | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01 |
Chemical Identifiers
7722-84-1 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |