missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, Fisher Chemical™
Supplier: Thermo Fisher Scientific H3124
Specifications
Hydrogen Peroxide | |
3% | |
Pass Test | |
0°C | |
H2O2 | |
1.00g/cm³ | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01 | |
USP | |
Pass Test | |
23.3mmHg at 30°C | |
Liquid |
Undesignated | |
7722-84-1,7732-18-5 | |
100°C | |
3% w/v | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
3.0 to 3.5% | |
3% w/v | |
Glass Bottle | |
4 L |
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention.
Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of