missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals

Used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

Supplier:  Thermo Scientific Chemicals 01232514

 View more versions of this product

Catalog No. AA1232514


Description

Description

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydrogen tetrachloroaurate(III) hydrate
Glass bottle
MFCD00149903
UN3260
Tetrachloroauric(III) acid,Chloroauric acid
RFWCEGCWRSNAGT-UHFFFAOYSA-K
hydrogen tetrachlorogoldtris(ylium)
339.79 (anhydrous)
Crystalline
27988-77-8
AuCl4H
25 g
Hygroscopic; Light sensitive
Soluble in water. Insoluble in ether.
[H+].Cl[Au+3](Cl)(Cl)Cl
339.77
99.9%

Chemical Identifiers

27988-77-8
MFCD00149903
hydrogen tetrachlorogoldtris(ylium)
[H+].Cl[Au+3](Cl)(Cl)Cl
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 240-948-4

RTECSNumber : MD5428000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only