Contec™ HydroKlean Solution
Sterile fill low endotoxin cleaner with hydrogen peroxide
$268.35 - $442.95
Catalog Number | Mfr. No. | Volume (Metric) | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Quantity | Price | Quantity | ||||
19-456-888
|
Contec™
SBT17HKLE |
0.5 L | 8/Cs. |
Case of 8 Each for $268.35
|
|
||||
19-456-889
|
Contec™
SBC170HKLE |
5 L | 2/Cs. |
Case of 2 Each for $442.95
|
|
||||
Description
Features:
- Blend of 6% hydrogen peroxide and water for injection in a 17 oz. and 34 oz. (1 L) bag in bottle with installed trigger spray and 170 oz. (5 L) capped container
- Ready to use and generates no Volatile Organic Compounds (VOCs)
- Hydrogen peroxide breaks down to water and oxygen in use, so it leaves little to no residue on a surface and is sutible for product contact areas
- Features a certified endotoxin level of <0.25 EU/ml making it ideal for use in product contact areas
- 0.2μm filtered and sterile filled into pre-irradiated containers under Grade A unidirectional airflow and bagged in a Grade A or ISO Class 5 cleanroom
- Bag in bottle system protects contents during use
- Certificates of analysis are available for each batch
- 6% hydrogen peroxide is not classed as corrosive and can be used safely in all areas of a cleanroom
- Nonflammable and ideal for use across large areas of a cleanroom or where sparks may be an issue
- Very low residue and eliminates the need for a residue removal stage with alcohol or water
- Double or triple bagged allowing for ease of entry into controlled environments
- Linear tear bags are easy to open even when wearing gloves
- Endotoxin level of less than 0.25 EU/mL
- Sterile filled into pre-irradiated containers in an ISO Class 5 cleanroom makes it suitable for use in all areas of the cleanroom including ISO Class 5 cabinets and RABS
- Double bagged or triple bagged in linear tear packaging facilitates transfer into isolators and RABS
- Trigger spray prevents bottle contents from being contaminated during use
- Not classified as corrosive can be used safely in all areas of the cleanroom
Specifications
Solution | |
Sterile |