missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Hydrol Pre-filled Containers, 10% NBF
Hydrol containers are made of polypropylene and are bulk packaged in corrugate trays. These are packaged in larger quantities and are ideal for larger labs with high throughput and have the ability to store large amounts of containers.
Supplier: Epredia™ LC0120
Specifications
Pre-filled Specimen Container | |
120 mL | |
CH2O | |
200/Cs. | |
C=O | |
30.03 |
Polypropylene | |
10% Formalin | |
MFCD00003274 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde |