missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroquinone, 99.5%, Thermo Scientific Chemicals

$43.56 - $63.57

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC219930050
SDS View Documents
Thermo Scientific Chemicals
219930050
5 g Glass Bottle
Each for $43.56
 
AC219930500
SDS View Documents
Thermo Scientific Chemicals
219930500
50 g Plastic bottle
Each for $44.31
 
AC219932500
SDS View Documents
Thermo Scientific Chemicals
219932500
250 g Plastic bottle
Each for $63.57
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-31-9
110.11
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
785
benzene-1,4-diol
C6H6O2
MFCD00002339
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
CHEBI:17594
C1=CC(=CC=C1O)O

Specifications

123-31-9
White
285.0°C to 287.0°C
Authentic
Glass Bottle
C6H4(OH)2
5 g
05,341; 14,249
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
benzene-1,4-diol
785
110.11
Needle-like Crystals or Crystalline Powder
170.0°C to 174.0°C
1.32
165°C
99.4% min. (HPLC)
C6H6O2
MFCD00002339
06,836
15,4845
Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol
C1=CC(=CC=C1O)O
110.11
CHEBI:17594
99.5%
Hydroquinone, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-617-8

RTECSNumber : MX3500000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only