missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroquinone bis(2-hydroxyethyl) ether, 95%, Thermo Scientific Chemicals

$61.25 - $483.52

Chemical Identifiers

CAS 104-38-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00002861
InChI Key WTPYFJNYAMXZJG-UHFFFAOYSA-N
Synonym hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
PubChem CID 66912
IUPAC Name 2-[4-(2-hydroxyethoxy)phenoxy]ethanol
SMILES C1=CC(=CC=C1OCCO)OCCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2403722
View Documents
Thermo Scientific Chemicals
B2403722
100 g
Each for $61.25
 
AAB2403736
View Documents
Thermo Scientific Chemicals
B2403736
500 g
Each for $134.75
 
AAB240370E
View Documents
Thermo Scientific Chemicals
B240370E
2500 g
Each for $483.52
 
Description

Description

Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

104-38-1
198.218
WTPYFJNYAMXZJG-UHFFFAOYSA-N
66912
C1=CC(=CC=C1OCCO)OCCO
C10H14O4
MFCD00002861
hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
2-[4-(2-hydroxyethoxy)phenoxy]ethanol

Specifications

104-38-1
C10H14O4
100 g
hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
WTPYFJNYAMXZJG-UHFFFAOYSA-N
2-[4-(2-hydroxyethoxy)phenoxy]ethanol
66912
95%
100°C to 102°C
MFCD00002861
Hygroscopic
Insoluble in water.
C1=CC(=CC=C1OCCO)OCCO
198.218
198.22
Hydroquinone bis(2-hydroxyethyl) ether
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-197-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only