missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HydroSpec™ Composit T2, Ricca Chemical

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

$453.68 - $453.68

Specifications

Specific Gravity 1
CAS 111-90-0,288-32-4,7446-09-5,7553-56-2
CAS Min % 60,10,5,1
CAS Max % 90,20,20,5
Packaging Amber Glass Bottle
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RK2100001C
View Documents
Ricca Chemical Company
RK2100001C
1 L
Each for $453.68
 
Description

Description

Specifications

Specifications

1
60,10,5,1
Amber Glass Bottle
UN3082
111-90-0,288-32-4,7446-09-5,7553-56-2
90,20,20,5
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'